Assay Details
Target Gene Details
Entrez Gene ID: | 208263 |
Gene Name: | torsin A interacting protein 1 |
Gene Aliases: |
LAP1, Lap1B |
Location: |
Chr.1:156004599-156036480 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tor1aip1 | NM_001160018.1 | NP_001153490.1 | ||
NM_001160019.1 | NP_001153491.1 | |||
NM_144791.2 | NP_659040.2 | |||
AB251963.1 | ||||
AK149727.1 | BAE29050.1 | |||
AK152751.1 | BAE31466.1 | |||
BC010841.1 | AAH10841.1 | |||
BQ891636.1 | ||||
CA465752.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |