Assay Details
Target Gene Details
Entrez Gene ID: | 56695 |
Gene Name: | paroxysmal nonkinesiogenic dyskinesia |
Gene Aliases: |
2210013N15Rik, 2810403H05Rik, AI854243, Brp17, MNCb-5687, MR-1, Tahccp2 |
Location: |
Chr.1:74284597-74353692 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnkd | NM_001039509.1 | NP_001034598.1 | ||
AK173125.1 | BAD32403.1 | |||
BC058945.1 | AAH58945.1 |
Target Gene Details
Entrez Gene ID: | 69660 |
Gene Name: | transmembrane BAX inhibitor motif containing 1 |
Gene Aliases: |
2310061B02Rik, AA960455, AU024746, C78899, RECS1, Tmbib1, mKIAA4161 |
Location: |
Chr.1:74288247-74304336 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmbim1 | NM_027154.5 | NP_081430.3 | ||
AB097685.1 | BAC43762.1 | |||
AK077689.1 | BAC36957.1 | |||
AK144110.1 | BAE25704.1 | |||
AK147297.1 | ||||
AK152864.1 | BAE31555.1 | |||
AK166926.1 | ||||
AK167286.1 | BAE39392.1 | |||
AK170625.1 | BAE41919.1 | |||
AK171102.1 | BAE42250.1 | |||
AK180422.1 | ||||
AK186797.1 | ||||
AK188714.1 | ||||
AK220296.1 | 5 | 982 | BAD90221.1 | |
BC004752.1 | AAH04752.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |