Assay Details
Target Gene Details
Entrez Gene ID: | 240690 |
Gene Name: | suppression of tumorigenicity 18 |
Gene Aliases: |
AV348974, Myt3, NZF-3, Nzf3, mKIAA0535 |
Location: |
Chr.1:6487231-6860940 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
St18 | NM_001244692.1 | NP_001231621.1 | ||
NM_001244693.1 | NP_001231622.1 | |||
NM_173868.2 | NP_776293.1 | |||
NR_045188.1 | ||||
NR_045189.1 | ||||
AB097467.1 | ||||
AJ536155.1 | CAD60089.1 | |||
AK032557.1 | ||||
AK122303.1 | BAC65585.1 | |||
AK133743.1 | BAE21817.1 | |||
AK137440.1 | BAE23351.1 | |||
AK141513.1 | BAE24709.1 | |||
AK147320.1 | BAE27844.1 | |||
AK147664.1 | BAE28057.1 | |||
BC118528.1 |
More Information
Set Membership: |
Intragenic Non-exonic |