Assay Details
Target Gene Details
Entrez Gene ID: | 70225 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 3 |
Gene Aliases: |
2310076N22Rik, 2510026K04Rik, Cyp10l |
Location: |
Chr.1:58430993-58445527 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppil3 | NM_001285826.1 | NP_001272755.1 | ||
NM_001285827.1 | NP_001272756.1 | |||
NM_027351.3 | NP_081627.1 | |||
NM_027374.3 | NP_081650.2 | |||
AF315803.1 | AAO32944.1 | |||
AK010207.1 | BAB26768.1 | |||
AK011000.1 | BAB27319.1 | |||
BC060993.1 | AAH60993.1 | |||
BC061645.1 | AAH61645.1 | |||
BY288296.1 | ||||
CJ166559.1 | ||||
CJ170065.1 | ||||
DV043111.1 |
More Information
Set Membership: |
Intragenic Non-exonic |