Assay Details
Target Gene Details
Entrez Gene ID: | 72823 |
Gene Name: | par-3 family cell polarity regulator beta |
Gene Aliases: |
1810008K04Rik, 2010002N16Rik, 2810455B10Rik, Als2cr19, PAR3B, PAR3L, PAR3beta |
Location: |
Chr.1:61638667-62642284 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pard3b | NM_001081050.2 | 14 | 2389 | NP_001074519.2 |
AK008055.1 | 14 | 2211 |