Assay Details
Target Gene Details
Entrez Gene ID: | 14768 |
Gene Name: | LanC (bacterial lantibiotic synthetase component C)-like 1 |
Gene Aliases: |
AW124738, Gpr69, Gpr69a, p40 |
Location: |
Chr.1:67000517-67038872 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lancl1 | NM_001190984.1 | NP_001177913.1 | ||
NM_001190985.1 | 1 | 93 | NP_001177914.1 | |
NM_021295.3 | NP_067270.1 | |||
AJ294535.1 | CAC69241.1 | |||
AK146120.1 | 1 | 85 | ||
BC058560.1 | AAH58560.1 | |||
Y11550.1 | CAA72314.1 | |||
Y16518.1 | 1 | 53 | CAA76269.1 |