Assay Details
Target Gene Details
Entrez Gene ID: | 98256 |
Gene Name: | kynurenine 3-monooxygenase (kynurenine 3-hydroxylase) |
Gene Aliases: |
AI046660 |
Location: |
Chr.1:175632193-175660853 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kmo | NM_133809.1 | NP_598570.1 | ||
AK158251.1 | ||||
AK171812.1 | ||||
BC014683.1 | AAH14683.1 |
More Information
Set Membership: |
Intragenic Non-exonic |