Assay Details
Target Gene Details
Entrez Gene ID: | 208263 |
Gene Name: | torsin A interacting protein 1 |
Gene Aliases: |
LAP1, Lap1B |
Location: |
Chr.1:156004599-156036480 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tor1aip1 | NM_001160018.1 | 1 | 450 | NP_001153490.1 |
NM_144791.2 | 1 | 450 | NP_659040.2 | |
AB251963.1 | 1 | 390 | ||
AK139709.1 | 1 | 119 | ||
AK149727.1 | 1 | 370 | BAE29050.1 | |
BC010841.1 | 1 | 265 | AAH10841.1 | |
BQ891636.1 | 1 | 45 |
Target Gene Details
Entrez Gene ID: | 240832 |
Gene Name: | torsin A interacting protein 2 |
Gene Aliases: |
1110020D10Rik, 15kDa, A130072J07, AA103493, AW060462, AW610675, C77739, Ifrg15, Lull1 |
Location: |
Chr.1:156035664-156068861 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tor1aip2 | NM_001160180.1 | NP_001153652.1 | ||
NM_001160182.1 | NP_001153654.1 | |||
NM_172843.4 | 1 | 368 | NP_766431.3 | |
AK038024.1 | 1 | 630 | BAC29923.1 | |
AK150117.1 | BAE29320.1 | |||
AK179038.1 | ||||
BU614106.1 | 1 | 368 | ||
BY254976.1 |