Assay Details
Target Gene Details
Entrez Gene ID: | 215303 |
Gene Name: | calcium/calmodulin-dependent protein kinase I gamma |
Gene Aliases: |
CLICK-III, CaMKIgamma |
Location: |
Chr.1:193346346-193370292 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Camk1g | NM_144817.2 | NP_659066.1 | ||
AF428262.1 | AAL28101.1 | |||
AY212936.1 | AAP29965.1 | |||
BC021840.1 | AAH21840.1 |
More Information
Set Membership: |
Intragenic Non-exonic |