Assay Details
Target Gene Details
Entrez Gene ID: | 12503 |
Gene Name: | CD247 antigen |
Gene Aliases: |
4930549J05Rik, A430104F18Rik, AW552088, Cd3, Cd3-eta, Cd3-zeta, Cd3h, Cd3z, Cd3zeta, T3z, Tcrk, Tcrz |
Location: |
Chr.1:165780357-165871153 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd247 | NM_001113391.2 | NP_001106862.1 | ||
NM_001113392.2 | NP_001106863.1 | |||
NM_001113393.2 | NP_001106864.2 | |||
NM_031162.4 | NP_112439.1 | |||
NR_103716.1 | ||||
AK017904.1 | BAB30997.1 | |||
AK088087.1 | ||||
AK138774.1 | BAE23774.1 | |||
BC052824.1 | AAH52824.1 | |||
J04967.1 | AAA50301.1 | |||
L03353.1 | AAA37401.1 | |||
M33158.1 | AAA37398.1 | |||
S51932.1 | AAB24559.1 | |||
U56258.1 | AAD11545.1 | |||
X84237.1 | CAA59015.1 |
More Information
Set Membership: |
Intragenic Non-exonic |