Assay Details
Target Gene Details
Entrez Gene ID: | 226527 |
Gene Name: | cDNA sequence BC026585 |
Gene Aliases: |
- |
Location: |
Chr.1:157458422-157490000 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BC026585 | NM_001033284.1 | NP_001028456.1 | ||
AK143913.1 | BAE25599.1 | |||
AK151310.1 | BAE30292.1 | |||
BC045198.1 |
More Information
Set Membership: |
Intragenic Non-exonic |