Assay Details
Target Gene Details
Entrez Gene ID: | 74104 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 6 |
Gene Aliases: |
1200005B17Rik |
Location: |
Chr.1:75171640-75180283 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcb6 | NM_023732.2 | NP_076221.1 | ||
AK004605.1 | BAB23404.1 | |||
AK040323.1 | 10 | 2449 | ||
AK151165.1 | BAE30168.1 | |||
AK164003.1 | ||||
AK168642.1 | BAE40501.1 | |||
AK200467.1 | ||||
AK200865.1 | ||||
AK208919.1 | ||||
AK212610.1 | ||||
AK218549.1 | ||||
BC006634.1 | AAH06634.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |