Assay Details
Target Gene Details
Entrez Gene ID: | 13508 |
Gene Name: | Down syndrome cell adhesion molecule |
Gene Aliases: |
4932410A21Rik |
Location: |
Chr.16:96587452-97170735 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dscam | NM_031174.4 | 29 | 5454 | NP_112451.1 |
AF315558.1 | 29 | 5099 | AAG28796.1 | |
AK134979.1 | ||||
AK141963.1 | 18 | 2521 | ||
AY005483.1 | 29 | 5454 | AAF99440.1 |
More Information
Set Membership: |
Intragenic Exonic |