Assay Details
Target Gene Details
Entrez Gene ID: | 56088 |
Gene Name: | proteasome (prosome, macropain) assembly chaperone 1 |
Gene Aliases: |
AW552102, Dscr2 |
Location: |
Chr.16:95979935-95990903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmg1 | NM_019537.2 | NP_062410.1 | ||
AJ238270.1 | CAB87195.1 | |||
AK042203.1 | BAC31197.1 | |||
AK088118.1 | BAC40157.1 | |||
BC032965.1 | AAH32965.1 | |||
BC060285.1 | AAH60285.1 |
More Information
Set Membership: |
Intragenic Non-exonic |