Assay Details
Target Gene Details
Entrez Gene ID: | 56176 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class P |
Gene Aliases: |
Dcrc, Dscr5 |
Location: |
Chr.16:94358763-94371015 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pigp | NM_001159617.1 | 1 | 206 | NP_001153089.1 |
NM_019543.3 | NP_062416.1 | |||
AF216309.1 | AAF32297.1 | |||
BC038252.1 | 1 | 217 | AAH38252.1 |
Target Gene Details
Entrez Gene ID: | 22129 |
Gene Name: | tetratricopeptide repeat domain 3 |
Gene Aliases: |
2610202A04Rik, AA409221, D16Ium21, D16Ium21e, TPRD, mKIAA4119 |
Location: |
Chr.16:94370739-94469222 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc3 | NM_009441.2 | 1 | 72 | NP_033467.2 |
AK053765.1 | 1 | 66 | BAC35513.1 | |
AK084201.1 | ||||
AK088273.1 | 1 | 73 | BAC40250.1 | |
AK089974.1 | BAC41023.1 | |||
AK165850.1 | 1 | 45 | BAE38412.1 | |
AK166120.1 | ||||
AK166164.1 | ||||
AK166298.1 | ||||
AK170087.1 | 1 | 63 | ||
AK220495.1 | BAD90293.1 | |||
BC019173.1 | ||||
BC049277.1 | 1 | 54 | ||
BC057207.1 | AAH57207.1 |
More Information
Set Membership: |
Intragenic Exonic |