Assay Details
Target Gene Details
Entrez Gene ID: | 15980 |
Gene Name: | interferon gamma receptor 2 |
Gene Aliases: |
Ifgr2, Ifgt |
Location: |
Chr.16:91547094-91564007 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ifngr2 | NM_008338.3 | 6 | 865 | NP_032364.1 |
AK137165.1 | 6 | 888 | BAE23260.1 | |
AK143214.1 | 6 | 880 | BAE25310.1 | |
AK165193.1 | 6 | 881 | BAE38071.1 | |
AK169907.1 | 6 | 886 | ||
BC055745.1 | 6 | 865 | AAH55745.1 | |
S69336.1 | 6 | 889 | AAB30165.1 |
More Information
Set Membership: |
Intragenic Exonic |