Assay Details
Target Gene Details
Entrez Gene ID: | 15976 |
Gene Name: | interferon (alpha and beta) receptor 2 |
Gene Aliases: |
AI747302, Ifnar-2 |
Location: |
Chr.16:91372215-91405589 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ifnar2 | NM_001110498.1 | 7 | 1040 | NP_001103968.1 |
NM_010509.2 | NP_034639.2 | |||
AF013274.1 | AAC53351.1 | |||
AF013486.1 | 7 | 912 | AAC53352.1 | |
AK003185.1 | 7 | 974 | BAB22628.1 | |
AK010952.1 | BAB27287.3 | |||
AK139169.1 | BAE23909.1 | |||
AK150386.1 | BAE29516.1 | |||
AK150597.1 | BAE29690.1 | |||
AK151474.1 | BAE30430.1 | |||
AK151493.1 | BAE30446.1 | |||
AK151777.1 | ||||
AK152878.1 | BAE31563.1 | |||
AK153317.1 | BAE31897.1 | |||
AK205320.1 | ||||
AK210331.1 | ||||
AK218073.1 | 1 | 54 | ||
BC003743.1 | 6 | 735 | AAH03743.1 | |
BC071225.1 | AAH71225.1 | |||
BE993108.1 | 1 | 148 | ||
Y09813.1 | 7 | 795 | CAA70943.1 | |
Y09864.1 | CAA70991.1 | |||
Y09865.1 | CAA70992.1 |
More Information
Set Membership: |
Intragenic Exonic |