Assay Details
Target Gene Details
Entrez Gene ID: | 23387 |
Gene Name: | SIK family kinase 3 |
Gene Aliases: |
L19, QSK, SIK-3 |
Location: |
Chr.11:116843402-117098453 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIK3 | NM_001281748.1 | NP_001268677.1 | ||
NM_001281749.1 | NP_001268678.1 | |||
NM_025164.4 | NP_079440.3 | |||
XM_005271481.2 | XP_005271538.1 | |||
XM_005271482.4 | XP_005271539.2 | |||
XM_005271484.3 | XP_005271541.2 | |||
XM_005271485.3 | XP_005271542.1 | |||
XM_011542721.2 | XP_011541023.1 | |||
XM_011542722.1 | XP_011541024.1 | |||
XM_011542723.2 | XP_011541025.1 | |||
XM_011542724.2 | XP_011541026.1 | |||
XM_011542725.2 | XP_011541027.1 | |||
XM_017017424.1 | XP_016872913.1 | |||
XM_017017425.1 | XP_016872914.1 | |||
XM_017017426.1 | XP_016872915.1 | |||
XM_017017427.1 | XP_016872916.1 | |||
AB209327.1 | BAD92564.1 | |||
BC063887.1 | AAH63887.1 | |||
BC128510.1 | ||||
BC128511.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv245e199 | Chr.11:116865114 - 116866642 on Build GRCh38 | Deletion | SIK3 |
esv24446 | Chr.11:116865124 - 116866382 on Build GRCh38 | Loss | SIK3 |
esv3548401 | Chr.11:116865103 - 116866479 on Build GRCh38 | Deletion | SIK3 |
esv3627837 | Chr.11:116865108 - 116866442 on Build GRCh38 | Loss | SIK3 |
nsv556454 | Chr.11:116865227 - 116867330 on Build GRCh38 | Loss | SIK3 |
nsv556451 | Chr.11:116863829 - 116865963 on Build GRCh38 | Loss | SIK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |