Assay Details
Target Gene Details
Entrez Gene ID: | 5478 |
Gene Name: | peptidylprolyl isomerase A |
Gene Aliases: |
CYPA, CYPH, HEL-S-69p |
Location: |
Chr.7:44796636-44803123 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPIA | NM_001300981.1 | NP_001287910.1 | ||
NM_021130.4 | NP_066953.1 | |||
AB451307.1 | ||||
AB451438.1 | ||||
AK130101.1 | ||||
AK290085.1 | ||||
AK290851.1 | ||||
AK293003.1 | ||||
AK297340.1 | ||||
AK316564.1 | ||||
AU100120.1 | ||||
BC093076.1 | AAH93076.1 | |||
BC137057.1 | ||||
BC137058.1 | ||||
BM806224.1 | ||||
BU786475.1 | ||||
CD014106.1 | ||||
CR456707.1 | CAG32988.1 | |||
Y00052.1 | CAA68264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830985 | Chr.7:44793475 - 44979588 on Build GRCh38 | Loss | MYO1G PURB MIR4657 PPIA H2AFV |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |