Assay Details
Target Gene Details
Entrez Gene ID: | 1124 |
Gene Name: | chimerin 2 |
Gene Aliases: |
ARHGAP3, BCH, CHN2-3, RHOGAP3 |
Location: |
Chr.7:29146547-29514335 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHN2 | NM_001293069.1 | NP_001279998.1 | ||
NM_001293070.1 | NP_001279999.1 | |||
NM_001293071.1 | NP_001280000.1 | |||
NM_001293072.1 | NP_001280001.1 | |||
NM_004067.3 | NP_004058.1 | |||
XM_011515105.2 | XP_011513407.1 | |||
XM_011515106.2 | XP_011513408.1 | |||
XM_011515107.2 | XP_011513409.1 | |||
XM_017011721.1 | XP_016867210.1 | |||
XM_017011722.1 | XP_016867211.1 | |||
AK293959.1 | ||||
AK294032.1 | ||||
AK294271.1 | ||||
AK313021.1 | ||||
BC112155.1 | AAI12156.1 | |||
DA096970.1 | ||||
DA105459.1 | ||||
DA190643.1 | ||||
EU732759.1 | ||||
EU732760.1 | ||||
EU732761.1 | ||||
EU732762.1 | ||||
EU732763.1 | ||||
EU732764.1 | ||||
EU732765.1 | ||||
EU732766.1 | ||||
GQ924106.1 | ||||
U28926.1 | AAA86528.1 |
Target Gene Details
Entrez Gene ID: | 102724484 |
Gene Name: | uncharacterized LOC102724484 |
Gene Aliases: |
- |
Location: |
Chr.7:29199040-29208970 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC102724484 | NR_120522.1 | |||
BU154957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3612685 | Chr.7:28997136 - 29209454 on Build GRCh38 | Loss | LOC101928168 CHN2 LOC100506497 CPVL LOC102724484 |
nsv1019174 | Chr.7:29163105 - 29499710 on Build GRCh38 | Gain | CHN2 LOC102724484 |
esv2761314 | Chr.7:29151010 - 29498723 on Build GRCh38 | Gain | CHN2 LOC102724484 |
dgv11248n54 | Chr.7:29169736 - 29497091 on Build GRCh38 | Gain | CHN2 LOC102724484 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |