Assay Details
Target Gene Details
Entrez Gene ID: | 5300 |
Gene Name: | peptidylprolyl cis/trans isomerase, NIMA-interacting 1 |
Gene Aliases: |
DOD, UBL5 |
Location: |
Chr.19:9835207-9849689 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIN1 | NM_006221.3 | NP_006212.1 | ||
NR_038422.2 | ||||
NR_038830.1 | ||||
XM_011528068.2 | XP_011526370.1 | |||
AF086512.1 | ||||
AK092970.1 | ||||
AK097558.1 | ||||
AK291074.1 | ||||
BC002899.1 | AAH02899.1 | |||
BC031971.1 | AAH31971.1 | |||
BG912418.1 | ||||
BI827520.1 | ||||
BI827672.1 | ||||
BT019331.1 | AAV38138.1 | |||
CD675162.1 | ||||
CR407654.1 | CAG28582.1 | |||
U49070.1 | AAC50492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520683 | Chr.19:9807553 - 9968417 on Build GRCh38 | Loss | PIN1 COL5A3 UBL5 OLFM2 FBXL12 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |