Assay Details
Target Gene Details
Entrez Gene ID: | 9229 |
Gene Name: | DLG associated protein 1 |
Gene Aliases: |
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 |
Location: |
Chr.18:3496032-4455307 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP1 | NM_001242761.1 | NP_001229690.1 | ||
NM_004746.3 | NP_004737.2 | |||
XM_005258171.2 | XP_005258228.1 | |||
XM_005258172.1 | XP_005258229.1 | |||
XM_011525770.2 | XP_011524072.1 | |||
XM_011525771.2 | XP_011524073.1 | |||
AK304101.1 | ||||
AK316099.1 |
Target Gene Details
Entrez Gene ID: | 284215 |
Gene Name: | DLGAP1 antisense RNA 5 |
Gene Aliases: |
- |
Location: |
Chr.18:4264602-4296000 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP1-AS5 | NR_036489.1 | |||
AK098180.1 | ||||
DA301744.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv184n21 | Chr.18:3526777 - 5930980 on Build GRCh38 | Gain | TMEM200C DLGAP1 MIR3976HG MIR6718 DLGAP1-AS3 LOC100286986 EPB41L3 ZBTB14 DLGAP1-AS4 DLGAP1-AS2 LOC105371967 LINC00667 MIR3976 LINC00526 DLGAP1-AS5 AKAIN1 DLGAP1-AS1 |
esv2656305 | Chr.18:4177454 - 4305905 on Build GRCh38 | Loss | DLGAP1 DLGAP1-AS5 |
dgv3315n100 | Chr.18:4249408 - 4311534 on Build GRCh38 | Gain | DLGAP1 DLGAP1-AS5 |
esv3893065 | Chr.18:3982600 - 4405472 on Build GRCh38 | Gain | DLGAP1 DLGAP1-AS4 DLGAP1-AS5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |