Assay Details
Target Gene Details
Entrez Gene ID: | 100128731 |
Gene Name: | oligosaccharyltransferase complex subunit 4, non-catalytic |
Gene Aliases: |
- |
Location: |
Chr.2:27070472-27071699 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OST4 | NM_001134693.1 | NP_001128165.1 | ||
AA772256.1 | ||||
AL049447.1 | ||||
BF697310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv6744n54 | Chr.2:26973431 - 27151141 on Build GRCh38 | Loss | OST4 EMILIN1 TCF23 TMEM214 LOC100129724 AGBL5-AS1 AGBL5 KHK PREB CGREF1 ABHD1 TRA-AGC8-1 PRR30 MAPRE3 |
nsv817673 | Chr.2:27037601 - 27163931 on Build GRCh38 | Gain | AGBL5-AS1 OST4 AGBL5 KHK EMILIN1 PREB CGREF1 ABHD1 TRA-AGC8-1 PRR30 TCF23 TMEM214 |
nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss | ATRAID OST4 EMILIN1 SLC30A3 TCF23 AGBL5 KHK SLC5A6 PREB CGREF1 CAD ABHD1 PRR30 |
nsv455974 | Chr.2:26989775 - 27128411 on Build GRCh38 | Loss | LOC100129724 AGBL5-AS1 OST4 AGBL5 KHK EMILIN1 CGREF1 ABHD1 TRA-AGC8-1 MAPRE3 TMEM214 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |