Assay Details
Target Gene Details
Entrez Gene ID: | 100128731 |
Gene Name: | oligosaccharyltransferase complex subunit 4, non-catalytic |
Gene Aliases: |
- |
Location: |
Chr.2:27070472-27071699 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OST4 | NM_001134693.1 | NP_001128165.1 | ||
AA772256.1 | ||||
AL049447.1 | ||||
BF697310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv6744n54 | Chr.2:26973431 - 27151141 on Build GRCh38 | Loss | TRA-AGC8-1 AGBL5-AS1 CGREF1 TMEM214 EMILIN1 AGBL5 PREB ABHD1 KHK MAPRE3 LOC100129724 PRR30 TCF23 OST4 |
nsv817673 | Chr.2:27037601 - 27163931 on Build GRCh38 | Gain | ABHD1 KHK TRA-AGC8-1 AGBL5-AS1 CGREF1 TMEM214 EMILIN1 AGBL5 PRR30 PREB TCF23 OST4 |
nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss | SLC30A3 CGREF1 EMILIN1 AGBL5 PREB ATRAID ABHD1 KHK SLC5A6 PRR30 CAD TCF23 OST4 |
nsv455974 | Chr.2:26989775 - 27128411 on Build GRCh38 | Loss | ABHD1 KHK MAPRE3 TRA-AGC8-1 AGBL5-AS1 CGREF1 LOC100129724 TMEM214 EMILIN1 AGBL5 OST4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |