Assay Details
Target Gene Details
Entrez Gene ID: | 123745 |
Gene Name: | phospholipase A2 group IVE |
Gene Aliases: |
- |
Location: |
Chr.15:41981582-42050703 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G4E | NM_001206670.1 | NP_001193599.1 | ||
XM_011521237.2 | XP_011519539.1 | |||
XM_011521238.1 | XP_011519540.1 | |||
XM_011521239.1 | XP_011519541.1 | |||
AK127558.1 | BAC87034.1 | |||
BC101584.1 | ||||
BC101612.1 | ||||
BX648482.1 |
Target Gene Details
Entrez Gene ID: | 101928388 |
Gene Name: | PLA2G4E antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:41972763-41999094 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G4E-AS1 | NR_120334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain | EHD4-AS1 LOC105370792 MAPKBP1 PLA2G4E-AS1 PLA2G4F SPTBN5 JMJD7 PLA2G4D PLA2G4E PLA2G4B EHD4 MIR4310 JMJD7-PLA2G4B |
nsv1038933 | Chr.15:41846258 - 42016889 on Build GRCh38 | Gain | EHD4-AS1 LOC105370792 SPTBN5 PLA2G4E-AS1 PLA2G4E PLA2G4B EHD4 MIR4310 JMJD7-PLA2G4B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |