Assay Details
Target Gene Details
Entrez Gene ID: | 6935 |
Gene Name: | zinc finger E-box binding homeobox 1 |
Gene Aliases: |
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A |
Location: |
Chr.10:31318495-31529814 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831824 | Chr.10:31290791 - 31340754 on Build GRCh38 | Loss | ZEB1 ZEB1-AS1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |