Assay Details
Target Gene Details
Entrez Gene ID: | 6935 |
Gene Name: | zinc finger E-box binding homeobox 1 |
Gene Aliases: |
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A |
Location: |
Chr.10:31318495-31529814 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZEB1 | NM_001323638.1 | 2 | 166 | NP_001310567.1 |
NM_001323641.1 | 1 | 210 | NP_001310570.1 | |
NM_001323642.1 | 1 | 210 | NP_001310571.1 | |
NM_001323643.1 | 2 | 166 | NP_001310572.1 | |
NM_001323644.1 | 1 | 210 | NP_001310573.1 | |
NM_001323645.1 | 1 | 210 | NP_001310574.1 | |
NM_001323646.1 | 1 | 210 | NP_001310575.1 | |
AK294775.1 | 1 | 175 | ||
AK309331.1 | 1 | 210 | ||
AK311372.1 | 1 | 156 | ||
BX647794.1 | 2 | 167 |
Target Gene Details
Entrez Gene ID: | 220930 |
Gene Name: | ZEB1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:31316528-31319095 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZEB1-AS1 | NR_024284.1 | 1 | 104 | |
AK296146.1 | ||||
BX446976.2 | ||||
DB121883.1 | 1 | 104 | ||
DB127940.1 | 1 | 66 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831824 | Chr.10:31290791 - 31340754 on Build GRCh38 | Loss | ZEB1-AS1 ZEB1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |