Assay Details
Target Gene Details
Entrez Gene ID: | 22839 |
Gene Name: | DLG associated protein 4 |
Gene Aliases: |
DAP-4, DAP4, DLP4, SAPAP-4, SAPAP4 |
Location: |
Chr.20:36306323-36528637 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP4 | NM_014902.5 | NP_055717.2 | ||
AB023181.1 | ||||
AL120584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066673 | Chr.20:35840156 - 36398197 on Build GRCh38 | Loss | DLGAP4 NORAD CNBD2 PHF20 EPB41L1 AAR2 SCAND1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |