Assay Details
Target Gene Details
Entrez Gene ID: | 101926987 |
Gene Name: | DLGAP4 antisense RNA 1 |
Gene Aliases: |
CCAT7 |
Location: |
Chr.20:36507702-36573275 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP4-AS1 | NR_109939.1 |
Target Gene Details
Entrez Gene ID: | 10398 |
Gene Name: | myosin light chain 9 |
Gene Aliases: |
LC20, MLC-2C, MLC2, MRLC1, MYRL2 |
Location: |
Chr.20:36541484-36549823 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYL9 | NM_006097.4 | NP_006088.2 | ||
NM_181526.2 | NP_852667.1 | |||
AF176042.1 | AAQ13653.1 | |||
AF176043.1 | AAQ13654.1 | |||
AK130879.1 | ||||
AL697699.1 | ||||
BC002648.2 | AAH02648.1 | |||
CR456843.1 | CAG33124.1 | |||
D82057.1 | BAB88917.1 | |||
J02854.1 | AAA59852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss | SLA2 TGIF2 DLGAP4-AS1 TGIF2-C20orf24 C20orf24 DLGAP4 MYL9 NDRG3 |
nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss | SOGA1 SLA2 TGIF2 DLGAP4-AS1 TGIF2-C20orf24 C20orf24 DSN1 DLGAP4 MYL9 NDRG3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |