Assay Details
Target Gene Details
Entrez Gene ID: | 79133 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 5 |
Gene Aliases: |
C20orf7, bA526K24.2, dJ842G6.1 |
Location: |
Chr.20:13785026-13821580 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFAF5 | NM_001039375.2 | NP_001034464.1 | ||
NM_024120.4 | NP_077025.2 | |||
NR_029377.1 | ||||
XM_006723620.3 | XP_006723683.1 | |||
XM_006723622.2 | XP_006723685.1 | |||
XM_006723623.2 | XP_006723686.1 | |||
XM_006723624.2 | XP_006723687.1 | |||
XM_011529342.2 | XP_011527644.1 | |||
XM_011529344.2 | XP_011527646.1 | |||
XM_017028061.1 | XP_016883550.1 | |||
AK091060.1 | ||||
AK289781.1 | ||||
BC005984.1 | AAH05984.1 | |||
BC073158.1 | AAH73158.1 | |||
BM993545.1 | ||||
CB156360.1 | ||||
DA289766.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1061303 | Chr.20:13414899 - 13830344 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)