Assay Details
Target Gene Details
Entrez Gene ID: | 8785 |
Gene Name: | matrilin 4 |
Gene Aliases: |
- |
Location: |
Chr.20:45293446-45308566 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MATN4 | NM_003833.4 | NP_003824.2 | ||
NM_030590.3 | NP_085080.1 | |||
NM_030592.3 | NP_085095.1 | |||
XM_005260597.1 | XP_005260654.1 | |||
XM_017028113.1 | XP_016883602.1 | |||
XM_017028114.1 | XP_016883603.1 | |||
XM_017028115.1 | XP_016883604.1 | |||
AJ007581.1 | CAA07569.1 | |||
AK027323.1 | ||||
AK074495.1 | ||||
AK074593.1 | ||||
AK074595.1 | BAC11081.1 | |||
AK074597.1 | BAC11083.1 | |||
BC131763.1 | ||||
BC141811.1 | ||||
BC151219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056815 | Chr.20:45257396 - 45314077 on Build GRCh38 | Loss | RBPJL MATN4 |
nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss | WFDC2 WFDC8 KCNK15 RBPJL SEMG2 MATN4 PI3 SLPI EPPIN-WFDC6 WFDC12 EPPIN STK4 STK4-AS1 SEMG1 SYS1-DBNDD2 SYS1 WFDC10A WFDC6 TOMM34 SDC4 DBNDD2 WISP2 SPINT3 TP53TG5 WFDC5 YWHAB PIGT PABPC1L MIR6812 KCNS1 KCNK15-AS1 RIMS4 WFDC9 |
nsv1057759 | Chr.20:45283246 - 45307625 on Build GRCh38 | Gain | RBPJL MATN4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |