Assay Details
Target Gene Details
Entrez Gene ID: | 90199 |
Gene Name: | WAP four-disulfide core domain 8 |
Gene Aliases: |
C20orf170, HEL-S-292, WAP8, dJ461P17.1 |
Location: |
Chr.20:45551152-45579326 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WFDC8 | NM_130896.2 | NP_570966.2 | ||
NM_181510.2 | NP_852611.2 | |||
XM_017028119.1 | XP_016883608.1 | |||
AF492015.1 | AAN70997.1 | |||
AF492016.1 | AAN70998.1 | |||
GQ472209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3645933 | Chr.20:45548287 - 45567209 on Build GRCh38 | Loss | WFDC8 |
nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss | TP53TG5 MIR6812 WFDC5 SEMG2 RBPJL WFDC2 SYS1 RIMS4 WFDC8 PABPC1L SLPI YWHAB PIGT SPINT3 STK4 EPPIN WFDC9 KCNS1 SEMG1 TOMM34 MATN4 EPPIN-WFDC6 KCNK15 SDC4 SYS1-DBNDD2 WFDC10A DBNDD2 WFDC6 PI3 KCNK15-AS1 STK4-AS1 WFDC12 WISP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |