Assay Details
Target Gene Details
Entrez Gene ID: | 149708 |
Gene Name: | WAP four-disulfide core domain 5 |
Gene Aliases: |
PRG5, WAP1, dJ211D12.5 |
Location: |
Chr.20:45109452-45116321 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WFDC5 | NM_145652.3 | NP_663627.1 | ||
XM_011528601.1 | XP_011526903.1 | |||
XM_011528602.1 | XP_011526904.1 | |||
AY038181.1 | AAK72468.1 | |||
AY358822.1 | AAQ89181.1 | |||
BC039173.1 | AAH39173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss | MIR6812 PABPC1L SLPI SEMG1 PI3 KCNK15-AS1 MATN4 WFDC9 RBPJL TP53TG5 KCNK15 TOMM34 SYS1-DBNDD2 EPPIN-WFDC6 SDC4 SPINT3 SEMG2 YWHAB WFDC2 STK4-AS1 WFDC5 DBNDD2 STK4 WFDC8 KCNS1 SYS1 WFDC10A EPPIN RIMS4 PIGT WFDC6 WISP2 WFDC12 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |