Assay Details
Target Gene Details
Entrez Gene ID: | 57419 |
Gene Name: | solute carrier family 24 member 3 |
Gene Aliases: |
NCKX3 |
Location: |
Chr.20:19212646-19722897 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC24A3 | NM_020689.3 | NP_065740.2 | ||
AF169257.2 | AAG12988.2 | |||
AF288087.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |