Assay Details
Target Gene Details
Entrez Gene ID: | 140901 |
Gene Name: | serine/threonine kinase 35 |
Gene Aliases: |
CLIK1, STK35L1 |
Location: |
Chr.20:2098035-2148555 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STK35 | NM_080836.3 | NP_543026.2 | ||
XM_011529174.2 | XP_011527476.1 | |||
AF195026.1 | AAL99353.1 | |||
AK302629.1 | ||||
AK314728.1 | ||||
BC017340.2 | AAH17340.2 | |||
BC111573.1 | AAI11574.1 | |||
BC140883.1 | ||||
BC140884.1 | ||||
GQ281297.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain | LOC727993 SNORD86 SNORD119 ZNF343 SNORD56 SNRPB LOC388780 SNORD57 SNORD110 EBF4 SNORA51 TGM6 MIR1292 TGM3 PDYN STK35 NOP56 IDH3B TMC2 |
nsv3258 | Chr.20:2091289 - 2124098 on Build GRCh38 | Insertion | STK35 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |