Assay Details
Target Gene Details
Entrez Gene ID: | 51230 |
Gene Name: | PHD finger protein 20 |
Gene Aliases: |
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP |
Location: |
Chr.20:35771999-35950377 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHF20 | NM_016436.4 | NP_057520.2 | ||
XM_011528843.1 | XP_011527145.1 | |||
XM_017027864.1 | XP_016883353.1 | |||
XM_017027868.1 | XP_016883357.1 | |||
XM_017027869.1 | XP_016883358.1 | |||
XM_017027870.1 | XP_016883359.1 | |||
XM_017027871.1 | XP_016883360.1 | |||
AF220416.1 | AAF34184.1 | |||
AF258787.1 | AAG49888.1 | |||
AK054597.1 | ||||
AK314503.1 | ||||
AY134747.1 | AAN08621.1 | |||
BC048210.1 | AAH48210.1 | |||
BC080598.1 | AAH80598.1 | |||
BC093405.1 | AAH93405.1 | |||
BC150178.1 | ||||
BG778940.1 | ||||
BM906582.1 | ||||
BX643875.1 | ||||
CD690482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056503 | Chr.20:35750468 - 35805624 on Build GRCh38 | Gain | PHF20 |
nsv526188 | Chr.20:35576541 - 36019079 on Build GRCh38 | Gain | SCAND1 PHF20 RBM12 FER1L4 SPAG4 CNBD2 ROMO1 CPNE1 NFS1 RBM39 |
dgv4303n100 | Chr.20:35730307 - 35805624 on Build GRCh38 | Gain | PHF20 RBM39 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |