Assay Details
Target Gene Details
Entrez Gene ID: | 140885 |
Gene Name: | signal regulatory protein alpha |
Gene Aliases: |
BIT, CD172A, MFR, MYD-1, P84, PTPNS1, SHPS1, SIRP |
Location: |
Chr.20:1894117-1939895 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIRPA | NM_001040022.1 | NP_001035111.1 | ||
NM_001040023.1 | NP_001035112.1 | |||
NM_080792.2 | NP_542970.1 | |||
XM_005260669.3 | XP_005260726.1 | |||
XM_005260670.3 | XP_005260727.1 | |||
XM_006723545.3 | XP_006723608.1 | |||
XM_011529173.2 | XP_011527475.1 | |||
AB023430.1 | BAA87929.1 | |||
AK290776.1 | ||||
AK298243.1 | ||||
AK312521.1 | ||||
BC026692.1 | AAH26692.1 | |||
BC033092.1 | AAH33092.1 | |||
BC038510.2 | AAH38510.1 | |||
BC075849.1 | AAH75849.1 | |||
D86043.1 | BAA12974.1 | |||
DN989886.1 | ||||
U06701.1 | ||||
Y10375.1 | CAA71403.1 | |||
Y11047.1 | CAA71944.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |