Assay Details
Target Gene Details
Entrez Gene ID: | 64849 |
Gene Name: | solute carrier family 13 member 3 |
Gene Aliases: |
NADC3, SDCT2 |
Location: |
Chr.20:46557823-46684485 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC13A3 | NM_001011554.2 | NP_001011554.1 | ||
NM_001193339.1 | NP_001180268.1 | |||
NM_001193340.1 | NP_001180269.1 | |||
NM_001193342.1 | NP_001180271.1 | |||
NM_022829.5 | NP_073740.2 | |||
AF154121.1 | AAF73251.1 | |||
AK056713.1 | BAB71262.1 | |||
AK096658.1 | BAC04834.1 | |||
AK123351.1 | ||||
AK293902.1 | ||||
AK295748.1 | ||||
AK303709.1 | ||||
AK310767.1 | ||||
AK310960.1 | ||||
AK315131.1 | ||||
AL442082.1 | CAC09447.2 | |||
AY072810.3 | AAL66762.1 | |||
BC035966.1 | AAH35966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833994 | Chr.20:46442116 - 46680210 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1061899 | Chr.20:46498425 - 47439490 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)