Assay Details
Target Gene Details
Entrez Gene ID: | 57325 |
Gene Name: | lysine acetyltransferase 14 |
Gene Aliases: |
ATAC2, CRP2BP, CSRP2BP, PRO1194, dJ717M23.1 |
Location: |
Chr.20:18142224-18188387 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KAT14 | NM_020536.4 | NP_065397.3 | ||
NR_028402.2 | ||||
AK023759.1 | BAB14669.1 | |||
AK297597.1 | ||||
AU140233.1 | ||||
BC007537.1 | AAH07537.1 | |||
BC009174.2 | AAH09174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833933 | Chr.20:18029399 - 18206602 on Build GRCh38 | Gain | OVOL2 PET117 KAT14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |