Assay Details
Target Gene Details
Entrez Gene ID: | 23315 |
Gene Name: | solute carrier family 9 member A8 |
Gene Aliases: |
NHE-8, NHE8 |
Location: |
Chr.20:49812713-49892242 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3646017 | Chr.20:49610386 - 49857106 on Build GRCh38 | Gain |
![]() ![]() |
esv3646034 | Chr.20:49788665 - 49826482 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)