Assay Details
Target Gene Details
Entrez Gene ID: | 57719 |
Gene Name: | anoctamin 8 |
Gene Aliases: |
KIAA1623, TMEM16H |
Location: |
Chr.19:17323223-17334834 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANO8 | NM_020959.2 | NP_066010.1 | ||
XM_017027048.1 | XP_016882537.1 | |||
AB046843.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470129 | Chr.19:17188084 - 17341718 on Build GRCh38 | Loss | ANO8 MYO9B NR2F6 USHBP1 OCEL1 GTPBP3 ABHD8 MRPL34 BABAM1 DDA1 ANKLE1 USE1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |