Assay Details
Target Gene Details
Entrez Gene ID: | 8425 |
Gene Name: | latent transforming growth factor beta binding protein 4 |
Gene Aliases: |
ARCL1C, LTBP-4, LTBP4L, LTBP4S |
Location: |
Chr.19:40592888-40629820 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LTBP4 | NM_001042544.1 | NP_001036009.1 | ||
NM_003573.2 | NP_003564.2 | |||
XM_011527376.2 | XP_011525678.1 | |||
XM_011527377.2 | XP_011525679.1 | |||
XM_011527378.2 | XP_011525680.1 | |||
XM_011527380.2 | XP_011525682.1 | |||
XM_011527381.2 | XP_011525683.1 | |||
XM_011527382.2 | XP_011525684.1 | |||
XM_011527383.2 | XP_011525685.1 | |||
XM_011527384.2 | XP_011525686.1 | |||
XM_011527385.2 | XP_011525687.1 | |||
XM_011527386.2 | XP_011525688.1 | |||
XM_011527387.1 | XP_011525689.1 | |||
XM_017027352.1 | XP_016882841.1 | |||
XM_017027353.1 | XP_016882842.1 | |||
XM_017027354.1 | XP_016882843.1 | |||
AF051345.2 | AAC39880.2 | |||
AK309484.1 | ||||
Y13622.1 | CAA73944.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |