Assay Details
Target Gene Details
Entrez Gene ID: | 84941 |
Gene Name: | hematopoietic SH2 domain containing |
Gene Aliases: |
ALX, HSH2 |
Location: |
Chr.19:16134028-16158575 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSH2D | NM_001291274.1 | NP_001278203.1 | ||
NM_032855.3 | NP_116244.1 | |||
NR_111903.1 | ||||
NR_111904.1 | ||||
AY319652.1 | AAQ81285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833767 | Chr.19:16121391 - 16347760 on Build GRCh38 | Loss | KLF2 FAM32A RAB8A CIB3 HSH2D AP1M1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |