Assay Details
Target Gene Details
Entrez Gene ID: | 22900 |
Gene Name: | caspase recruitment domain family member 8 |
Gene Aliases: |
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN |
Location: |
Chr.19:48203148-48256269 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CARD8 | XM_017026485.1 | XP_016881974.1 | ||
XM_017026490.1 | XP_016881979.1 | |||
AF143869.1 | ||||
AK302327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | FAM83E PTOV1-AS1 SIGLEC16 RRAS CABP5 SNAR-C1 SNAR-B2 SNAR-A12 TBC1D17 MIR4324 C19orf68 SPHK2 SNORD35B CGB1 SNORD34 LOC105447645 NOSIP BAX VRK3 SNAR-C3 PRMT1 CCDC114 RPL13A NUP62 SNAR-A6 GRWD1 LOC100287477 RASIP1 KCNJ14 TEAD2 SNAR-A11 CARD8 FCGRT MAMSTR KDELR1 SULT2B1 DKKL1 SNAR-G2 GRIN2D SNORD33 EMP3 NUCB1 MIR4749 GFY FTL ATF5 SNAR-B1 SNAR-A8 CYTH2 NTN5 RPL18 LIG1 IZUMO1 SLC6A16 SNORD32A ZNF114 RCN3 PIH1D1 GYS1 DBP KCNA7 PRRG2 SNAR-A7 LOC101928295 LOC101059948 TMEM143 SNAR-C4 SNAR-A3 PTH2 FGF21 RPS11 MED25 SNAR-A14 HRC ADM5 LOC105372430 BSPH1 ZNF473 CGB3 SNAR-G1 SNAR-C5 FLT3LG CGB2 HSD17B14 SNAR-A13 SNAR-A1 TRPM4 IL4I1 LHB PTOV1-AS2 TSKS SLC17A7 CD37 PTOV1 IRF3 PRR12 SNAR-A10 C19orf73 SYNGR4 FUT2 SNRNP70 PNKP PLA2G4C-AS1 SNAR-A9 SNORD35A CARD8-AS1 FUT1 BCL2L12 MIR5088 PLA2G4C CCDC155 CA11 CGB5 LMTK3 AP2A1 SNAR-A4 AKT1S1 MIR6798 MIR6800 BCAT2 CGB8 SNAR-C2 SNAR-D NUCB1-AS1 SNAR-A2 MIR150 PPFIA3 FUZ ELSPBP1 DHDH PPP1R15A RUVBL2 SNAR-A5 SIGLEC11 SEC1P FLJ26850 TULP2 SCAF1 CGB7 NTF4 MIR4750 ALDH16A1 CPT1C MIR4751 SPACA4 PLEKHA4 MIR6799 LIN7B |
esv34181 | Chr.19:48009377 - 48416682 on Build GRCh38 | Loss | CCDC114 ELSPBP1 EMP3 PLA2G4C-AS1 CABP5 CARD8-AS1 CARD8 PLA2G4C ZNF114 C19orf68 KDELR1 LIG1 TMEM143 SYNGR4 GRIN2D |
nsv523716 | Chr.19:48198962 - 48208219 on Build GRCh38 | Loss | CARD8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |