Assay Details
Target Gene Details
Entrez Gene ID: | 3171 |
Gene Name: | forkhead box A3 |
Gene Aliases: |
FKHH3, HNF3G, TCF3G |
Location: |
Chr.19:45864260-45873797 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXA3 | NM_004497.2 | NP_004488.2 | ||
AK223473.1 | BAD97193.1 | |||
BC016024.1 | AAH16024.1 | |||
BT006720.1 | AAP35366.1 | |||
L12141.1 | AAA58477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510770 | Chr.19:45841909 - 45921434 on Build GRCh38 | Deletion | SYMPK MYPOP IRF2BP1 NANOS2 FOXA3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |