Assay Details
Target Gene Details
Entrez Gene ID: | 10892 |
Gene Name: | MALT1 paracaspase |
Gene Aliases: |
IMD12, MLT, MLT1, PCASP1 |
Location: |
Chr.18:58671386-58753806 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MALT1 | NM_006785.3 | NP_006776.1 | ||
NM_173844.2 | NP_776216.1 | |||
XM_011525794.1 | XP_011524096.1 | |||
AB026118.1 | BAA83099.1 | |||
AF130356.2 | AAD38507.2 | |||
AF316597.1 | AAG38589.1 | |||
AK291386.1 | ||||
BC030143.2 | AAH30143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv577050 | Chr.18:58664431 - 58732147 on Build GRCh38 | Loss | LOC101927322 MALT1 LOC102724745 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |