Assay Details
Target Gene Details
Entrez Gene ID: | 79944 |
Gene Name: | L-2-hydroxyglutarate dehydrogenase |
Gene Aliases: |
C14orf160, L2HGA |
Location: |
Chr.14:50242434-50312235 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L2HGDH | NM_024884.2 | NP_079160.1 | ||
XM_005268075.4 | XP_005268132.1 | |||
XM_011537166.2 | XP_011535468.1 | |||
XM_011537167.2 | XP_011535469.1 | |||
XM_011537168.2 | XP_011535470.1 | |||
XM_017021655.1 | XP_016877144.1 | |||
XM_017021656.1 | XP_016877145.1 | |||
XM_017021657.1 | XP_016877146.1 | |||
AK022680.1 | BAB14174.1 | |||
AK292627.1 | ||||
AY757363.1 | AAV52330.1 | |||
BC006117.1 | AAH06117.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3634475 | Chr.14:50257216 - 50265757 on Build GRCh38 | Loss |
![]() |
nsv564835 | Chr.14:50024482 - 50439753 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)