Assay Details
Target Gene Details
Entrez Gene ID: | 394 |
Gene Name: | Rho GTPase activating protein 5 |
Gene Aliases: |
GFI2, RhoGAP5, p190-B, p190BRhoGAP |
Location: |
Chr.14:32076152-32159728 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGAP5 | NM_001030055.1 | NP_001025226.1 | ||
NM_001173.2 | NP_001164.2 | |||
XM_005267635.3 | XP_005267692.1 | |||
XM_005267636.3 | XP_005267693.1 | |||
XM_017021287.1 | XP_016876776.1 | |||
XM_017021288.1 | XP_016876777.1 | |||
AB209751.1 | BAD92988.1 | |||
AY316523.1 | AAP83192.1 | |||
BC050059.1 | ||||
BC075799.1 | AAH75799.1 | |||
BC129928.1 | ||||
BC129929.1 | ||||
CA945100.1 | ||||
U17032.1 | AAA95963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832764 | Chr.14:32095262 - 32254007 on Build GRCh38 | Gain | RNU6-7 ARHGAP5 RNU6-8 |
nsv564182 | Chr.14:31641313 - 32115131 on Build GRCh38 | Gain | ARHGAP5 NUBPL ARHGAP5-AS1 |
esv3584226 | Chr.14:32090990 - 32298443 on Build GRCh38 | Gain | RNU6-7 ARHGAP5 RNU6-8 |
nsv1046526 | Chr.14:31641331 - 32098220 on Build GRCh38 | Gain | ARHGAP5 NUBPL ARHGAP5-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |