Assay Details
Target Gene Details
Entrez Gene ID: | 729665 |
Gene Name: | coiled-coil domain containing 175 |
Gene Aliases: |
C14orf38, c14_5395 |
Location: |
Chr.14:59504212-59577276 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC175 | NM_001164399.1 | NP_001157871.1 | ||
XM_011537120.2 | XP_011535422.1 | |||
XM_011537122.2 | XP_011535424.1 | |||
XM_011537124.2 | XP_011535426.1 |