Assay Details
Target Gene Details
Entrez Gene ID: | 9628 |
Gene Name: | regulator of G-protein signaling 6 |
Gene Aliases: |
GAP |
Location: |
Chr.14:71932100-72632478 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGS6 | NM_001204416.2 | NP_001191345.1 | ||
NM_001204417.2 | NP_001191346.1 | |||
NM_001204418.2 | NP_001191347.1 | |||
NM_001204419.2 | NP_001191348.1 | |||
NM_001204420.2 | NP_001191349.1 | |||
NM_001204421.2 | NP_001191350.1 | |||
NM_001204422.2 | NP_001191351.1 | |||
NM_001204423.1 | NP_001191352.1 | |||
NM_001204424.1 | NP_001191353.1 | |||
NM_004296.6 | NP_004287.3 | |||
NR_135235.1 | ||||
XM_017021818.1 | XP_016877307.1 | |||
XM_017021819.1 | XP_016877308.1 | |||
XM_017021824.1 | XP_016877313.1 | |||
XM_017021829.1 | XP_016877318.1 | |||
AB209462.1 | BAD92699.1 | |||
AF073920.2 | AAC26049.2 | |||
AK294486.1 | ||||
AK294895.1 | ||||
AK313481.1 | ||||
BC104877.1 | AAI04878.1 | |||
BC113572.1 | ||||
DC317315.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |